Methyl laurate ≥98.0%

Supplier: TCI America

L0015-500ML L0015-25ML
TCL0015-500MLEA 202.97 CAD
TCL0015-500ML TCL0015-025ML
Methyl laurate ≥98.0%
Methyl laurate

CAS Number: 111-82-0
MDL Number: MFCD00008966
Molecular Formula: C13H26O2
Molecular Weight: 214.35
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 262
Melting point (°C): 5
Flash Point (°C): 134
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Lauric Acid Methyl Ester

Formula: C₁₃H₂₆O₂
Boiling Pt: 262 °C
Melting Pt: 5 °C
Density: 0.87 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f2130080c044337def023381daa82c6a11942a&vid=07f2130080c044337def023381daa82c6a11942a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR