Ethyl laurate ≥99.0%

Supplier: TCI America

Synonyms: Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester

L0013-25ML L0013-500ML
TCL0013-025MLEA 42.66 CAD
TCL0013-025ML TCL0013-500ML
Ethyl laurate ≥99.0%
Ethyl laurate

CAS Number: 106-33-2
MDL Number: MFCD00015065
Molecular Formula: C14H28O2
Molecular Weight: 228.38
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 275
Melting point (°C): -10
Flash Point (°C): 101
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Lauric Acid Ethyl Ester

Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 269
Melting Pt: -2
Density: 0.86 (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00015065
CAS Number: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08fdf82f709e3ba6f22296d45b977ac115b9cc05&vid=08fdf82f709e3ba6f22296d45b977ac115b9cc05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR