Lauric acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: Dodecanoic acid, Dodecylic acid

L0011-25G L0011-500G
TCL0011-025GEA 32.15 CAD
TCL0011-025G TCL0011-500G
Lauric acid ≥98.0%
Lauric acid

CAS Number: 143-07-7
MDL Number: MFCD00002736
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 298
Melting point (°C): 45
Flash Point (°C): 145
Lambda max.: 207 nm (MeOH)


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Pressure & Heat Sensitive Recording Materials, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Dodecanoic Acid

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296
Melting Pt: 45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5ef9d55108db0639b8795cd21f9664b09caed24&vid=6925feffc25737ccc153e1117bdc98408d3ea96e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR