Methyl butyrylacetate ≥96.0%

Supplier: TCI America

Synonyms: Methyl 3-oxohexanoate

K0037-500ML K0037-25ML
TCK0037-500MLEA 363.4 CAD
TCK0037-500ML TCK0037-025ML
Methyl butyrylacetate ≥96.0%
Methyl butyrylacetate

CAS Number: 30414-54-1
MDL Number: MFCD00040995
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 90
Specific Gravity (20/20): 1.02


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Butyrylacetic Acid Methyl Ester, 3-Ketohexanoic Acid Methyl Ester, Methyl Butyrylacetate, 3-Oxohexanoic Acid Methyl Ester

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 85 °C
Density: 1.02 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00040995
CAS Number: 30414-54-1
EINECS: 250-185-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=610b988711b84c145f1efbd35ac3dd6d4b1ff9fa&vid=610b988711b84c145f1efbd35ac3dd6d4b1ff9fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR