Methyl propionylacetate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

K0035-25G K0035-500G K0035-100G
TCK0035-025GEA 79.32 CAD
TCK0035-025G TCK0035-500G TCK0035-100G
Methyl propionylacetate ≥98.0%
Methyl propionylacetate

CAS Number: 30414-53-0
MDL Number: MFCD00011705
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 71
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 3-Ketovaleric Acid Methyl Ester, Methyl 3-Oxopentanoate, Methyl Propionylacetate, 3-Oxovaleric Acid Methyl Ester, Propionylacetic Acid Methyl Ester

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 74 °C
Density: 1.05 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00011705
CAS Number: 30414-53-0
EINECS: 250-184-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9776634625841668d8cd7185bba962be632f49&vid=eb9776634625841668d8cd7185bba962be632f49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR