Ethyl propionylacetate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate

K0031-5G K0031-25G
TCK0031-005GEA 50.56 CAD
TCK0031-005G TCK0031-25G
Ethyl propionylacetate ≥97.0%
Ethyl propionylacetate

CAS Number: 4949-44-4
MDL Number: MFCD00009317
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Flash Point (°C): 77
Specific Gravity (20/20): 1.01


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Bioscience, Lipids
Other Product Names / Synonyms: Ethyl 3-Oxopentanoate, Ethyl Propionylacetate, 3-Ketovaleric Acid Ethyl Ester, 3-Oxovaleric Acid Ethyl Ester, Propionylacetic Acid Ethyl Ester

Formula: C₇H₁₂O₃
Boiling Pt: 191 °C
Density: 1.01 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00009317
CAS Number: 4949-44-4
EINECS: 225-593-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5620c40816ecf30a2d43f8d312ef30233e8081f7&vid=5620c40816ecf30a2d43f8d312ef30233e8081f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR