Ethyl butyrylacetate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

K0030-25ML K0030-500ML
TCK0030-025MLEA 34.76 CAD
TCK0030-025ML TCK0030-500ML
Ethyl butyrylacetate ≥95.0%
Ethyl butyrylacetate

CAS Number: 3249-68-1
MDL Number: MFCD00009401
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Melting point (°C): -44
Flash Point (°C): 78
Specific Gravity (20/20): 0.99


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Bioscience, Lipids
Other Product Names / Synonyms: Ethyl Butyrylacetate, Ethyl 3-Ketohexanoate, 3-Ketohexanoic Acid Ethyl Ester, 3-Oxohexanoic Acid Ethyl Ester

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 95 °C
Melting Pt: –44 °C
Density: 0.99 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00009401
CAS Number: 3249-68-1
EINECS: 221-835-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3105e110095fecdc1b581ca3cc807e70a284c48&vid=f3105e110095fecdc1b581ca3cc807e70a284c48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR