Methyl-2-iodobenzoate ≥98.0%

Supplier: TCI America

I0586-25G I0586-250G
TCI0586-25GEA 98.96 CAD
TCI0586-25G TCI0586-250G
Methyl-2-iodobenzoate ≥98.0%
Methyl 2-iodobenzoate

CAS Number: 610-97-9
MDL Number: MFCD00016351
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 274
Flash Point (°C): 110
Specific Gravity (20/20): 1.79


Other Product Names / Synonyms: 2-Iodobenzoic Acid Methyl Ester

Formula: C₈H₇IO₂
Boiling Pt: 274 °C
Density: 1.79 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00016351
CAS Number: 610-97-9
EINECS: 210-243-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21f861f07991784ed1a79b9cc3240b42e9e93000&vid=21f861f07991784ed1a79b9cc3240b42e9e93000&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR