3-Iodopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0568-25G I0568-5G
TCI0568-025GEA 373.51 CAD
TCI0568-025G TCI0568-005G
3-Iodopyridine ≥98.0% (by GC, titration analysis)
3-Iodopyridine

CAS Number: 1120-90-7
MDL Number: MFCD00023553
Molecular Formula: C5H4IN
Molecular Weight: 205.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 92
Melting point (°C): 55

Formula: C₅H₄IN
Boiling Pt: 92 °C
Melting Pt: 55 °C
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d84926817656e8567fdca4cbd1b8fa18f0ab1a82&vid=d84926817656e8567fdca4cbd1b8fa18f0ab1a82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR