4-Iodo-m-xylene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,4-Dimethyliodobenzene, 1-Iodo-2,4-dimethylbenzene, 2,4-Dimethyl-1-iodobenzene, 1,3-Dimethyl-4-iodobenzene

I0561-25G I0561-5G
TCI0561-025GEA 91.64 CAD
TCI0561-025G TCI0561-5G
4-Iodo-m-xylene ≥98.0%
4-Iodo-m-xylene

CAS Number: 4214-28-2
MDL Number: MFCD00013706
Molecular Formula: C8H9I
Molecular Weight: 232.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Flash Point (°C): 110
Specific Gravity (20/20): 1.63


Other Product Names / Synonyms: 1-Iodo-2,4-dimethylbenzene

Formula: C₈H₉I
Boiling Pt: 233 °C
Density: 1.63 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013706
CAS Number: 4214-28-2
EINECS: 224-141-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=263c07852d01ae51b40e86998d5565817f1c2ce4&vid=624b9737eb3aaef2d57a2bf420d6f2bc1ad26701&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR