Isopropyltriphenylphosphonium iodide ≥98.0%

Supplier: TCI America

I0552-25G I0552-5G
TCI0552-25GEA 189.6 CAD
TCI0552-25G TCI0552-005G
Isopropyltriphenylphosphonium iodide ≥98.0%
Isopropyltriphenylphosphonium iodide

CAS Number: 24470-78-8
MDL Number: MFCD00031595
Molecular Formula: C21H22IP
Molecular Weight: 432.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199

Formula: C₂₁H₂₂IP
MW: 432.29 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00031595
CAS Number: 24470-78-8
EINECS: 246-281-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c7d12dd3fe07d5118fd8843a6d6e678adc8cb96&vid=8c7d12dd3fe07d5118fd8843a6d6e678adc8cb96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR