α,α,α-Trifluoro-4-iodotoluene ≥97.0% stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

I0550-25G I0550-5G
TCI0550-25GEA 285.98 CAD
TCI0550-25G TCI0550-005G
α,α,α-Trifluoro-4-iodotoluene ≥97.0% stabilized
α,α,α-Trifluoro-4-iodotoluene

(stabilized with Copper chip)
CAS Number: 455-13-0
MDL Number: MFCD00039398
Molecular Formula: C7H4F3I
Molecular Weight: 272.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 68
Specific Gravity (20/20): 1.88


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Iodo-4-(trifluoromethyl)benzene, 4-Iodo-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₄F₃I
Boiling Pt: 185 °C
Melting Pt: 17 °C
Density: 1.88 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00039398
CAS Number: 455-13-0
EINECS: 207-234-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7102b9cfd73b7a82015b1c30ef285bc8b12a2218&vid=eb1dc4f7841e33f2b68a3454dea99f68de4d1466&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR