5-Iodo-m-xylene ≥98.0% stabilised

Supplier: TCI America
Warning

Synonyms: 1-Iodo-3,5-dimethylbenzene, 3,5-Dimethyl-1-iodobenzene, 1,3-Dimethyl-5-iodobenzene, 3,5-Dimethyliodobenzene

Stabilization: Stabilised with Stabilised

I0548-5G I0548-25G
TCI0548-005GEA 96.38 CAD
TCI0548-005G TCI0548-25G
5-Iodo-m-xylene ≥98.0% stabilised
5-Iodo-m-xylene

CAS Number: 22445-41-6
MDL Number: MFCD00060659
Molecular Formula: C8H9I
Molecular Weight: 232.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 113
Specific Gravity (20/20): 1.61


Other Product Names / Synonyms: 3,5-Dimethyliodobenzene, 1-Iodo-3,5-dimethylbenzene

Formula: C₈H₉I
Boiling Pt: 229 °C
Density: 1.61 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060659
CAS Number: 22445-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e5f0f0b7a2659c8f3f7096d8a84c7c2f54c5bbf&vid=7e5f0f0b7a2659c8f3f7096d8a84c7c2f54c5bbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR