3-Iodo-o-xylene ≥97.0%

Supplier: TCI America
Warning

Synonyms: 1,2-Dimethyl-3-iodobenzene, 2,3-Dimethyl-1-iodobenzene, 1-Iodo-2,3-dimethylbenzene, 2,3-Dimethyliodobenzene

I0547-5G I0547-25G
TCI0547-005GEA 145.36 CAD
TCI0547-005G TCI0547-025G
3-Iodo-o-xylene ≥97.0%
3-Iodo-o-xylene

CAS Number: 31599-60-7
MDL Number: MFCD00041437
Molecular Formula: C8H9I
Molecular Weight: 232.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 110
Specific Gravity (20/20): 1.65


Other Product Names / Synonyms: 2,3-Dimethyliodobenzene, 1-Iodo-2,3-dimethylbenzene

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 231 °C
Density: 1.65 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041437
CAS Number: 31599-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c62e02e6f923045798a01fc31ecd050a0a69407&vid=2c62e02e6f923045798a01fc31ecd050a0a69407&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR