Methyl indole-5-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 1H-indole-5-carboxylate

I0545-1G I0545-5G
TCI0545-001GEA 48.98 CAD
TCI0545-001G TCI0545-5G
Methyl indole-5-carboxylate ≥98.0%
Methyl indole-5-carboxylate

CAS Number: 1011-65-0
MDL Number: MFCD00153023
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126


Other Product Names / Synonyms: Indole-5-carboxylic Acid Methyl Ester

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00153023
CAS Number: 1011-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5023bd16d6bf825682a9bac227ba73d2f071d5c9&vid=5023bd16d6bf825682a9bac227ba73d2f071d5c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR