Ethyl-3-iodobenzoate ≥98.0%

Supplier: TCI America

Synonyms: 3-Iodobenzoic acid ethyl ester

I0536-5G I0536-25G
TCI0536-005GEA 64.78 CAD
TCI0536-005G TCI0536-025G
Ethyl-3-iodobenzoate ≥98.0%
Ethyl 3-iodobenzoate

CAS Number: 58313-23-8
MDL Number: MFCD00044769
Molecular Formula: C9H9IO2
Molecular Weight: 276.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 110
Specific Gravity (20/20): 1.66


Other Product Names / Synonyms: 3-Iodobenzoic Acid Ethyl Ester

Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 161 °C
Density: 1.66 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00044769
CAS Number: 58313-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64e4d11eba2e2df0cb2a51c7d906706900ff4efb&vid=64e4d11eba2e2df0cb2a51c7d906706900ff4efb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR