4-Iodophenetole ≥98.0%

Supplier: TCI America

Synonyms: 4-Iodophenyl ethyl ether, 1-Ethoxy-4-iodobenzene, 1-Iodo-4-ethoxybenzene

I0525-25G I0525-5G
TCI0525-025GEA 135.88 CAD
TCI0525-025G TCI0525-5G
4-Iodophenetole ≥98.0%
4-Iodophenetole

CAS Number: 699-08-1
MDL Number: MFCD00061124
Molecular Formula: C8H9IO
Molecular Weight: 248.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 26


Other Product Names / Synonyms: 4-Ethoxyiodobenzene

Formula: C₈H₉IO
MW: 248.06 g/mol
MDL Number: MFCD00061124
CAS Number: 699-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba25b9f216449f0a9b6fa9fa2c4c272fe872e2b5&vid=ba25b9f216449f0a9b6fa9fa2c4c272fe872e2b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR