3-(Hydroxymethyl)indole ≥95.0% (by total nitrogen basis)

Supplier: TCI America

I0496-5G I0496-25G
TCI0496-5GEA 64.78 CAD
TCI0496-5G TCI0496-025G
3-(Hydroxymethyl)indole ≥95.0% (by total nitrogen basis)
3-(Hydroxymethyl)indole

CAS Number: 700-06-1
MDL Number: MFCD00005632
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Bioscience, Antioxidants
Other Product Names / Synonyms: 3-(Hydroxymethyl)indole, Indole-3-carbinol

Formula: C₉H₉NO
MDL Number: MFCD00005632
CAS Number: 700-06-1
EINECS: 211-836-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32adee6338a022600954052326beb972e80b8139&vid=32adee6338a022600954052326beb972e80b8139&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR