3-Iodophenol ≥98.0%

Supplier: TCI America
Warning

I0495-10G I0495-25G
TCI0495-010GEA 134.3 CAD
TCI0495-010G TCI0495-25G
3-Iodophenol ≥98.0%
3-Iodophenol

CAS Number: 626-02-8
MDL Number: MFCD00002261
Molecular Formula: C6H5IO
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 41
Flash Point (°C): 113

Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 190 °C
Melting Pt: 41 °C
Flash Pt: 113 °C
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2
UN: 3335
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2922dabe926fc9bf2a9fd1de8ccc2444724ec8fe&vid=b57552b7a02e8a887837c03a445d4647a6a67d86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR