1-Iododecane ≥96.0% stabilised

Supplier: TCI America
Warning

Synonyms: Decane, 1-iodo-, Decyl iodide

Stabilization: Stabilised with Stabilised

I0494-25G I0494-250G
TCI0494-025GEA 85.54 CAD
TCI0494-025G TCI0494-250G
1-Iododecane ≥96.0% stabilised
1-Iododecane

(stabilized with Copper chip)
CAS Number: 2050-77-3
MDL Number: MFCD00001085
Molecular Formula: C10H21I
Molecular Weight: 268.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 110
Specific Gravity (20/20): 1.26


Other Product Names / Synonyms: Decyl Iodide

Formula: C₁₀H₂₁I
Boiling Pt: 132 °C
Melting Pt: –16.3 °C
Density: 1.26 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001085
CAS Number: 2050-77-3
EINECS: 218-104-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5da403a20df291949a8e190f416d7eddc593369&vid=c1569cf257611bf62233757fbb98dff45336ab22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR