Methyl indole-3-carboxylate ≥98.0%

Supplier: TCI America

I0491-500G I0491-25G
TCI0491-500GEA 562.48 CAD
TCI0491-500G TCI0491-025G
Methyl indole-3-carboxylate ≥98.0%
Methyl indole-3-carboxylate

CAS Number: 942-24-5
MDL Number: MFCD00189407
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 149


Other Product Names / Synonyms: Indole-3-carboxylic Acid Methyl Ester

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00189407
CAS Number: 942-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3891f15692390a17db6bc01fd41ebc590eb4f57&vid=a3891f15692390a17db6bc01fd41ebc590eb4f57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR