2-Iodomesitylene ≥98.0%

Supplier: TCI America

Synonyms: 2,4,6-Trimethyliodobenzene, 1-Iodo-2,4,6-trimethylbenzene, 2-Iodo-1,3,5-trimethylbenzene, Iodomesitylene

I0478-25G I0478-5G
TCI0478-025GEA 99.54 CAD
TCI0478-025G TCI0478-5G
2-Iodomesitylene ≥98.0%
2-Iodomesitylene

CAS Number: 4028-63-1
MDL Number: MFCD00013707
Molecular Formula: C9H11I
Molecular Weight: 246.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 31
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Iodomesitylene, 2-Iodo-1,3,5-trimethylbenzene

Formula: C₉H₁₁I
MW: 246.09 g/mol
Boiling Pt: 250 °C
Melting Pt: 31 °C
MDL Number: MFCD00013707
CAS Number: 4028-63-1
EINECS: 223-709-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dd0c518f5b6f40a491579f46c0f2db096bceb5d&vid=1dd0c518f5b6f40a491579f46c0f2db096bceb5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR