5-Iodovanillin ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

I0427-25G I0427-5G
TCI0427-025GEA 138.27 CAD
TCI0427-025G TCI0427-005G
5-Iodovanillin ≥98.0%
5-Iodovanillin

CAS Number: 5438-36-8
MDL Number: MFCD00006941
Molecular Formula: C8H7IO3
Molecular Weight: 278.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

Formula: C₈H₇IO₃
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 182 °C
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c25a3237dae39d113417e0e28992f1cb729cb3b&vid=74fd583ff37240841ae3b46752ffb3db2ce6902c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR