(S)-(-)-Indoline-2-carboxylic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
I0395-5G
I0395-1G
TCI0395-005GEA
132.25
CAD
TCI0395-005G
TCI0395-1G
(S)-(-)-Indoline-2-carboxylic acid ≥95.0% (by titrimetric analysis)
(S)-(-)-Indoline-2-carboxylic acid
CAS Number: 79815-20-6
MDL Number: MFCD00070578
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Specific rotation [a]20/D: -116 deg (C=1, 2mol/L HCl)
Formula:
C₉H₉NO₂ |
MDL Number:
MFCD00070578 CAS Number: 79815-20-6 EINECS: 410-860-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...