3-Iodothiophene ≥95.0%

Supplier: TCI America

I0380-5G I0380-1G
TCI0380-005GEA 94.8 CAD
TCI0380-005G TCI0380-001G
3-Iodothiophene ≥95.0%
3-Iodothiophene

CAS Number: 10486-61-0
MDL Number: MFCD00037901
Molecular Formula: C4H3IS
Molecular Weight: 210.03
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Melting point (°C): -13
Flash Point (°C): 78
Specific Gravity (20/20): 2.06


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Thienyl Iodide

Formula: C₄H₃IS
Boiling Pt: 75 °C
Melting Pt: –13 °C
Density: 2.06 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037901
CAS Number: 10486-61-0
EINECS: 234-009-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9555b2248ad5aea455d3427148c4d32e4c627b13&vid=9555b2248ad5aea455d3427148c4d32e4c627b13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR