3-Iodoanisole ≥98.0%

Supplier: TCI America
Warning

I0379-25G I0379-5G
TCI0379-025GEA 140.62 CAD
TCI0379-025G TCI0379-5G
3-Iodoanisole ≥98.0%
3-Iodoanisole

CAS Number: 766-85-8
MDL Number: MFCD00001048
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 110
Specific Gravity (20/20): 1.77

Formula: C₇H₇IO
Boiling Pt: 245 °C
Density: 1.77 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001048
CAS Number: 766-85-8
EINECS: 212-173-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=592f529101319160f9f86cc3fc573c06ce5d2e0d&vid=592f529101319160f9f86cc3fc573c06ce5d2e0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR