4-Iodoanisole ≥99.0%
Supplier: TCI America
I0378-25G
I0378-100G
TCI0378-025GEA
47.87
CAD
TCI0378-025G
TCI0378-100G
4-Iodoanisole ≥99.0%
4-Iodoanisole
CAS Number: 696-62-8
MDL Number: MFCD00001056
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 51
Flash Point (°C): 110
Formula:
C₇H₇IO Boiling Pt: 240 °C Melting Pt: 51 °C Density: 1 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001056 CAS Number: 696-62-8 EINECS: 211-798-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...