(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol ≥97.0%

Supplier: TCI America

Synonyms: (+)-2,3-O-Isopropylidene-L-threitol

I0376-1G I0376-5G
TCI0376-001GEA 117.38 CAD
TCI0376-001G TCI0376-005G
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol ≥97.0%
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol

CAS Number: 50622-09-8
MDL Number: MFCD00063761
Molecular Formula: C7H14O4
Molecular Weight: 162.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110
Specific rotation [a]20/D: 3.9 deg (C=5, Acetone)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: (4S,5S)-(+)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane, (4S,5S)-(+)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol

Formula: C₇H₁₄O₄
MW: 162.19 g/mol
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00063761
CAS Number: 50622-09-8
EINECS: 256-658-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a53c3877246b84cc0c245623d1b3a2e478519155&vid=a53c3877246b84cc0c245623d1b3a2e478519155&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR