(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol ≥98.0%
Supplier: TCI America
CAS Number: 73346-74-4
MDL Number: MFCD00009761
Molecular Formula: C7H14O4
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 50
Flash Point (°C): 110
Specific rotation [a]20/D: -4 deg (C=5, Acetone)
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: (4R,5R)-(-)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane, (4R,5R)-(-)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol
Formula:
C₇H₁₄O₄ Boiling Pt: 96 °C Melting Pt: 50 °C Flash Pt: 110 °C |
MDL Number:
MFCD00009761 CAS Number: 73346-74-4 EINECS: 277-391-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...