2-Iodothiophene ≥98.0%
Supplier: TCI America
I0374-25G
I0374-100G
TCI0374-025GEA
188.02
CAD
TCI0374-025G
TCI0374-100G
2-Iodothiophene ≥98.0%
2-Iodothiophene
CAS Number: 3437-95-4
MDL Number: MFCD00005424
Molecular Formula: C4H3IS
Molecular Weight: 210.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Melting point (°C): -40
Flash Point (°C): 71
Specific Gravity (20/20): 2.06
Other Product Names / Synonyms: 2-Thienyl Iodide
Formula:
C₄H₃IS MW: 210.04 g/mol Boiling Pt: 182 °C Melting Pt: –40 °C Density: 2.06 (20 °C) Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005424 CAS Number: 3437-95-4 EINECS: 222-342-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...