4-Isopropylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0371-25ML I0371-500ML I0371-100ML
TCI0371-025MLEA 61.07 CAD
TCI0371-025ML TCI0371-500ML TCI0371-100ML
4-Isopropylaniline ≥98.0% (by GC, titration analysis)
4-Isopropylaniline

CAS Number: 99-88-7
MDL Number: MFCD00007900
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 224
Flash Point (°C): 116
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: 4-Aminocumene, p-Cumidine

Formula: C₉H₁₃N
Boiling Pt: 224 °C
Melting Pt: –63 °C
Density: 0.95 (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7
EINECS: 202-797-2
UN: 1760
ADR: 8,III
Merck Index: 13,02647
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89bc49521ffbecdf544c4dd952110e1a4bd991a2&vid=859af7e2f6b06fd754b41010fb3a512162e5779b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR