Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
I0366-5G
I0366-25G
TCI0366-005GEA
109.02
CAD
TCI0366-005G
TCI0366-025G
Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis)
Tetrabutylammonium chloride
[Reagent for Ion-Pair Chromatography]
CAS Number: 1112-67-0
MDL Number: MFCD00011635
Molecular Formula: C16H36ClN
Molecular Weight: 277.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 70
Flash Point (°C): 113
Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Acidic Samples
Other Product Names / Synonyms: IPC-TBA-Cl
Formula:
C₁₆H₃₆ClN Melting Pt: 70 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00011635 CAS Number: 1112-67-0 EINECS: 214-195-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...