Tetrabutylammonium bromide ≥99.0%

Supplier: TCI America

I0365-25G I0365-500G I0365-100G
TCI0365-025GEA 66.64 CAD
TCI0365-025G TCI0365-500G TCI0365-100G
Tetrabutylammonium bromide ≥99.0%
Tetrabutylammonium bromide

[Reagent for Ion-Pair Chromatography]
CAS Number: 1643-19-2
MDL Number: MFCD00011633
Molecular Formula: C16H36BrN
Molecular Weight: 322.38
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 103


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Acidic Samples
Other Product Names / Synonyms: IPC-TBA-Br

Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=687210e083cca81d851fd2d8ea8d8ed27134b482&vid=a2355fc23cb47a6641cfde8d06206edfab1d1a03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR