Sodium dodecyl sulfate (SDS) ≥97.0%

Supplier: TCI America
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

I0352-500G I0352-25G I0352-100G
TCI0352-500GEA 263.39 CAD
TCI0352-500G TCI0352-025G TCI0352-100G
Sodium dodecyl sulfate (SDS) ≥97.0%
Sodium dodecyl sulphate (SDS)

[Reagent for Ion-Pair Chromatography]
CAS Number: 151-21-3
MDL Number: MFCD00036175
Molecular Formula: C12H26O4S
Molecular Weight: 288.38
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 207
Flash Point (°C): 145


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-SDS, Laurylsulfuric Acid Sodium Salt, Dodecylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70133d1a576563b95828e0ed37231b08216ed2af&vid=0596d7502d61df67d5e9e3d5cf2392e080493266&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR