Sodium 1-decanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0348-5G I0348-25G
TCI0348-005GEA 57.88 CAD
TCI0348-005G TCI0348-025G
Sodium 1-decanesulfonate ≥98.0% (by titrimetric analysis)
Sodium 1-decanesulphonate

[Reagent for Ion-Pair Chromatography]
CAS Number: 13419-61-9
MDL Number: MFCD00007526
Molecular Formula: C10H22O3S
Molecular Weight: 244.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: 1-Decanesulfonic Acid Sodium Salt, IPC-ALKS-10

Formula: C₁₀H₂₁NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e693f9752dbb02932ec1fb51c73644095ce94387&vid=4fa29a49b6f90148ae955b8f5b2cdd2e21f76be3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR