1-Heptanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0345-5G I0345-25G I0345-100G
TCI0345-005GEA 63.2 CAD
TCI0345-005G TCI0345-025G TCI0345-100G
1-Heptanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1-Heptanesulphonic acid sodium salt

[Reagent for Ion-Pair Chromatography]
CAS Number: 22767-50-6
MDL Number: MFCD00007543
Molecular Formula: C7H16O3S
Molecular Weight: 202.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: 1-Heptanesulfonic Acid Sodium Salt, IPC-ALKS-7

Formula: H₃C(CH₂)₅CH₂SO₃Na
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f8712ae7500f81700dae7a1c7f8f4e40eed2fcd&vid=dc8c883888b9eee8b7e5384ad3393125028f1d8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR