1-Pentanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0343-25G I0343-5G I0343-100G
TCI0343-025GEA 188.11 CAD
TCI0343-025G TCI0343-005G TCI0343-100G
1-Pentanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1-Pentanesulphonic acid sodium salt

[Reagent for Ion-Pair Chromatography]
CAS Number: 22767-49-3
MDL Number: MFCD00007541
Molecular Formula: C5H12O3S
Molecular Weight: 174.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-ALKS-5, 1-Pentanesulfonic Acid Sodium Salt

Formula: C₅H₁₁O₃SNa
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d840d652ad09ac81b2a6396c594b1c259e2c79&vid=dca2905e8eaa2531e8dfd202d2b1ca647ea34032&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR