Indole-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0332-25G I0332-5G
TCI0332-025GEA 60.04 CAD
TCI0332-025G TCI0332-5G
Indole-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Indole-2-carboxylic acid

CAS Number: 1477-50-5
MDL Number: MFCD00005611
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 205

Formula: C₉H₇NO₂
Melting Pt: 205 °C
MDL Number: MFCD00005611
CAS Number: 1477-50-5
EINECS: 216-030-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc376222c44735ba68dab0a541954477ce06f291&vid=bc376222c44735ba68dab0a541954477ce06f291&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR