2-Isobutyl-3-methoxypyrazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

I0331-1ML I0331-5ML
TCI0331-001MLEA 91.64 CAD
TCI0331-001ML TCI0331-005ML
2-Isobutyl-3-methoxypyrazine ≥97.0% (by GC, titration analysis)
2-Isobutyl-3-methoxypyrazine

CAS Number: 24683-00-9
MDL Number: MFCD00006128
Molecular Formula: C9H14N2O
Molecular Weight: 166.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 80
Specific Gravity (20/20): 1.00

Formula: C₉H₁₄N₂O
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 107 °C
Density: 1.00 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00006128
CAS Number: 24683-00-9
EINECS: 246-402-1

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR