IPTG (isopropyl-β-D-thiogalactopyranoside) 98,0

Supplier: TCI America

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

I0328-1G I0328-5G
TCI0328-001GEA 108.01 CAD
TCI0328-001G TCI0328-005G
IPTG (isopropyl-β-D-thiogalactopyranoside) 98,0
IPTG (Isopropyl-β-D-thiogalactopyranoside)

CAS Number: 367-93-1
MDL Number: MFCD00063273
Molecular Formula: C9H18O5S
Molecular Weight: 238.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 113
Specific rotation [a]20/D: -32 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Thioglycosides, Glycosides, Galactose
Other Product Names / Synonyms: Isopropyl-beta-D-thiogalactopyranoside

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402
Melting Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71785053db2dfaa4317d40cc6d577803a0bb75ea&vid=71785053db2dfaa4317d40cc6d577803a0bb75ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR