1-Iodooctane ≥97.0% stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

I0321-25G I0321-500G I0321-100G
TCI0321-025GEA 74.26 CAD
TCI0321-025G TCI0321-500G TCI0321-100G
1-Iodooctane ≥97.0% stabilized
1-Iodooctane

(stabilized with Copper chip)
CAS Number: 629-27-6
MDL Number: MFCD00001105
Molecular Formula: C8H17I
Molecular Weight: 240.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 87
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: n-Octyl Iodide

Formula: C₈H₁₇I
Boiling Pt: 226 °C
Melting Pt: –46 °C
Density: 1.33 (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00001105
CAS Number: 629-27-6
EINECS: 211-084-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79df6f7fd083658f8d32df0a89aaa07d545ce021&vid=795dd93cde09225b2efab436c9f8251124a7a274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR