1-Iodoheptane ≥98.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

I0320-25G I0320-500G
TCI0320-025GEA 55.3 CAD
TCI0320-025G TCI0320-500G
1-Iodoheptane ≥98.0% stabilized
1-Iodoheptane

(stabilized with Copper chip)
CAS Number: 4282-40-0
MDL Number: MFCD00001104
Molecular Formula: C7H15I
Molecular Weight: 226.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 204
Melting point (°C): -48
Flash Point (°C): 78
Specific Gravity (20/20): 1.38


Other Product Names / Synonyms: Heptyl Iodide

Formula: C₇H₁₅I
Boiling Pt: 204 °C
Melting Pt: –48 °C
Density: 1.38 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001104
CAS Number: 4282-40-0
EINECS: 224-285-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2abde8a66d7ff6986e641a8fcf39a3ec0d0648d2&vid=992cdd0e2fe1f646d7528034aefa6020269c56de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR