2-Isopropylaniline ≥98.0%

Supplier: TCI America
Warning

I0318-25ML I0318-250ML
TCI0318-25MLEA 41.08 CAD
TCI0318-25ML TCI0318-250ML
2-Isopropylaniline ≥98.0%
2-Isopropylaniline

CAS Number: 643-28-7
MDL Number: MFCD00007720
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 96
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 2-Aminocumene, o-Cumidine

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 104 °C
Density: 0.97 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00007720
CAS Number: 643-28-7
EINECS: 211-397-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92f6a2e28d01d564dbe21c8f69b2e58e93b94861&vid=8bb72379fef1fc5fd2637f4b55fedddc8ad93230&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR