2-Isopropylimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0313-25G I0313-500G
TCI0313-025GEA 35.34 CAD
TCI0313-025G TCI0313-500G
2-Isopropylimidazole ≥98.0% (by GC, titration analysis)
2-Isopropylimidazole

CAS Number: 36947-68-9
MDL Number: MFCD00014486
Molecular Formula: C6H10N2
Molecular Weight: 110.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 132

Formula: C₆H₁₀N₂
Melting Pt: 132 °C
MDL Number: MFCD00014486
CAS Number: 36947-68-9
EINECS: 253-286-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e44f7dab75562a3e67ce976631d02e8f65e69cd0&vid=e44f7dab75562a3e67ce976631d02e8f65e69cd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR