3-Hydroxy-p-anisic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I0295-25G I0295-5G
TCI0295-010GEA 149.67 CAD
TCI0295-010G TCI0295-25G TCI0295-5G
3-Hydroxy-p-anisic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-p-anisic acid

CAS Number: 645-08-9
MDL Number: MFCD00002507
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 253


Other Product Names / Synonyms: 3-Hydroxy-4-methoxybenzoic Acid

Formula: C₈H₈O₄
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 253 °C
MDL Number: MFCD00002507
CAS Number: 645-08-9
EINECS: 211-430-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28ac79d1c9a5c8f4d68bbf0515f25cccc4dad411&vid=a5030d37c75fa42e114442d6a40f397a06f2ebe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR