Imidazole ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Glyoxaline1H-ImidazoleGlyoxalamine

I0288-25G I0288-100G
TCI0288-100GEA 1141.58 CAD
TCI0288-25G TCI0288-100G
Imidazole ≥99.0% (by GC, titration analysis)
Imidazole

[for Buffer]
CAS Number: 288-32-4
MDL Number: MFCD00005183
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 90
Flash Point (°C): 145

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 256 °C
Melting Pt: 90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 145 °C
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 2923
ADR: 8,III
Merck Index: 13,04935
https://ca.vwr.com/stibo/web/std.lang.all/20/96/28972096.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR