Indazole ≥99.0%

Supplier: TCI America

Synonyms: 1H-Indazole

I0278-1G I0278-5G
TCI0278-001GEA 51.44 CAD
TCI0278-001G TCI0278-005G
Indazole ≥99.0%
Indazole

CAS Number: 271-44-3
MDL Number: MFCD00005691
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 147


Other Product Names / Synonyms: 1,2-Benzodiazole, 1,2-Benzopyrazole

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 270
Melting Pt: 147 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00005691
CAS Number: 271-44-3
EINECS: 205-978-4
Merck Index: 13,04961
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c55c87bf69c708b411cba23ef768f62c372b86c&vid=5c55c87bf69c708b411cba23ef768f62c372b86c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR