4-Isopropylbenzyl alcohol ≥98.0%

Supplier: TCI America
Warning

Synonyms: (4-Isopropylphenyl)methanol

I0272-100ML I0272-25ML
TCI0272-100MLEA 99.54 CAD
TCI0272-100ML TCI0272-025ML
4-Isopropylbenzyl alcohol ≥98.0%
4-Isopropylbenzyl alcohol

CAS Number: 536-60-7
MDL Number: MFCD00004663
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Flash Point (°C): 113
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Cumic Alcohol, Cuminyl Alcohol

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 249
Melting Pt: 28
Density: 0.98 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004663
CAS Number: 536-60-7
EINECS: 208-640-4
Merck Index: 13,02646
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d243b0b0263cdd853210373ed6e6b4cf91d7372e&vid=d243b0b0263cdd853210373ed6e6b4cf91d7372e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR