Diethyl isopropylmalonate ≥98.0%
Supplier: TCI America
I0252-500ML
I0252-25ML
I0252-100ML
TCI0252-500MLEA
603.56
CAD
TCI0252-500ML
TCI0252-025ML
TCI0252-100ML
Diethyl isopropylmalonate ≥98.0%
Diethyl isopropylmalonate
CAS Number: 759-36-4
MDL Number: MFCD00040491
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 92
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Isopropylmalonic Acid Diethyl Ester
Formula:
C₁₀H₁₈O₄ Boiling Pt: 214 °C Density: 0.99 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00040491 CAS Number: 759-36-4 EINECS: 212-068-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...