4-Iodotoluene ≥99.0%

Supplier: TCI America
Warning

I0218-25G I0218-500G I0218-100G
TCI0218-025GEA 109.02 CAD
TCI0218-025G TCI0218-500G TCI0218-100G
4-Iodotoluene ≥99.0%
4-Iodotoluene

CAS Number: 624-31-7
MDL Number: MFCD00001059
Molecular Formula: C7H7I
Molecular Weight: 218.04
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 212
Flash Point (°C): 90
Freezing point (°C): 36

Formula: C₇H₇I
Boiling Pt: 212 °C
Melting Pt: 33…35 °C
Density: 1.68 g/cm³ (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7
EINECS: 210-841-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbaf4b78f4735e6c110e8c400a4167161f41e112&vid=9933bb546727ceb451bb694466c8b4d6f4159763&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR